Assay Details
Gene Symbol: LY6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284001.1 | NP_001270930.1 | 1 - 2 | 204 | 58 | |
XM_015454754.1 | XP_015310240.1 | 2 - 3 | 209 | 58 | ||
GenBank mRNA | AB170991.1 | - | 1 - 2 | 204 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
