Assay Details
| Gene Symbol: NUDT21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284772.1 | NP_001271701.1 | 2 - 3 | 431 | 71 | |
| GenBank mRNA | AB172174.1 | - | 2 - 3 | 431 | 71 | |
More Information
Set Membership: |
|

NUDT21
Genomic Map