Assay Details
Gene Symbol: SLC3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285242.2 | NP_001272171.2 | 820 | 56 | ||
NM_001354865.1 | NP_001341794.1 | 996 | 56 | |||
GenBank mRNA | AB169510.1 | - | 820 | 56 | ||
AB170184.1 | - | 820 | 56 |

More Information
Set Membership: |
![]() ![]() |
