Assay Details
| Gene Symbol: WNT9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283113.1 | NP_001270042.1 | 1 - 2 | 189 | 113 | |
| GenBank mRNA | AB172917.1 | - | 1 - 2 | 189 | 113 | |
More Information
Set Membership: |
|

WNT9A
Genomic Map