Assay Details
| Gene Symbol: WNT9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283113.1 | NP_001270042.1 | 3 - 4 | 712 | 86 | |
| XM_015435229.1 | XP_015290715.1 | 2 - 3 | 4600 | 86 | ||
| XM_015435552.1 | XP_015291038.1 | 3 - 4 | 4078 | 86 | ||
| GenBank mRNA | AB172917.1 | - | 3 - 4 | 712 | 86 | |
More Information
Set Membership: |
|

WNT9A
Genomic Map