Assay Details
| Gene Symbol: SGMS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283255.1 | NP_001270184.1 | 5 - 6 | 1308 | 79 | |
| XM_015450530.1 | XP_015306016.1 | 6 - 7 | 1445 | 79 | ||
| XM_015450531.1 | XP_015306017.1 | 6 - 7 | 1424 | 79 | ||
| XM_015450532.1 | XP_015306018.1 | 6 - 7 | 1427 | 79 | ||
| XM_015450533.1 | XP_015306019.1 | 6 - 7 | 1467 | 79 | ||
| XM_015450534.1 | XP_015306020.1 | 6 - 7 | 1464 | 79 | ||
| XM_015450535.1 | XP_015306021.1 | 5 - 6 | 1317 | 79 | ||
| XM_015450536.1 | XP_015306022.1 | 5 - 6 | 1357 | 79 | ||
| XM_015450537.1 | XP_015306023.1 | 5 - 6 | 1495 | 79 | ||
| XM_015450538.1 | XP_015306024.1 | 4 - 5 | 1675 | 79 | ||
| GenBank mRNA | AB168962.1 | - | 5 - 6 | 1308 | 79 | |
More Information
Set Membership: |
|

SGMS2
Genomic Map