Assay Details
Gene Symbol: POLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283289.1 | NP_001270218.1 | 6 - 7 | 1253 | 69 | |
XM_015455868.1 | XP_015311354.1 | 7 - 8 | 1401 | 69 | ||
XM_015455869.1 | XP_015311355.1 | 7 - 8 | 1398 | 69 | ||
XM_015455870.1 | XP_015311356.1 | 7 - 8 | 1357 | 69 | ||
XM_015455871.1 | XP_015311357.1 | 7 - 8 | 1377 | 69 | ||
XM_015455872.1 | XP_015311358.1 | 6 - 7 | 1227 | 69 | ||
XM_015455873.1 | XP_015311359.1 | 7 - 8 | 1125 | 69 | ||
XM_015455874.1 | XP_015311360.1 | 7 - 8 | 1401 | 69 | ||
XM_015455877.1 | XP_015311363.1 | 4 - 5 | 625 | 69 | ||
XM_015455878.1 | XP_015311364.1 | 4 - 5 | 622 | 69 | ||
XM_015455879.1 | XP_015311365.1 | 4 - 5 | 532 | 69 | ||
XM_015455880.1 | XP_015311366.1 | 4 - 5 | 601 | 69 | ||
GenBank mRNA | AB179386.1 | - | 6 - 7 | 1253 | 69 | |
HM486795.1 | - | 6 - 7 | 1194 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
