Assay Details
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283527.1 | NP_001270456.1 | 2 - 3 | 124 | 76 | |
XM_015434110.1 | XP_015289596.1 | 3 - 4 | 224 | 76 | ||
GenBank mRNA | AB072017.1 | - | 1 - 2 | 64 | 76 | |
AB169517.1 | - | 2 - 3 | 148 | 76 | ||
AB170328.1 | - | 2 - 3 | 134 | 76 | ||
AB171356.1 | - | 2 - 3 | 124 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
