Assay Details
Gene Symbol: C3H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283568.1 | NP_001270497.1 | 4 - 5 | 539 | 121 | |
XM_005549952.2 | XP_005550009.1 | 4 - 5 | 767 | 121 | ||
XM_005549953.2 | XP_005550010.1 | 4 - 5 | 767 | 121 | ||
GenBank mRNA | AB169092.1 | - | 4 - 5 | 539 | 121 |

More Information
Set Membership: |
![]() ![]() |
