Assay Details
Gene Symbol: C3H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283568.1 | NP_001270497.1 | 7 - 8 | 840 | 93 | |
GenBank mRNA | AB169092.1 | - | 7 - 8 | 840 | 93 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C3H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283568.1 | NP_001270497.1 | 7 - 8 | 840 | 93 | |
GenBank mRNA | AB169092.1 | - | 7 - 8 | 840 | 93 |