Assay Details
Gene Symbol: C3H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283568.1 | NP_001270497.1 | 9 - 10 | 1102 | 76 | |
GenBank mRNA | AB169092.1 | - | 9 - 10 | 1102 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
