Assay Details
Gene Symbol: TNFAIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283816.1 | NP_001270745.1 | 2 - 3 | 573 | 66 | |
XM_015448380.1 | XP_015303866.1 | 2 - 3 | 718 | 66 | ||
XM_015448381.1 | XP_015303867.1 | 2 - 3 | 565 | 66 | ||
XM_015448382.1 | XP_015303868.1 | 2 - 3 | 1581 | 66 | ||
XM_015448383.1 | XP_015303869.1 | 2 - 3 | 479 | 66 | ||
GenBank mRNA | AB168334.1 | - | 2 - 3 | 573 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
