Assay Details
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283987.1 | NP_001270916.1 | 3 - 4 | 235 | 75 | |
XM_005542088.2 | XP_005542145.1 | 3 - 4 | 273 | 75 | ||
XM_015451341.1 | XP_015306827.1 | 3 - 4 | 312 | 75 | ||
XM_015451348.1 | XP_015306834.1 | 3 - 4 | 276 | 75 | ||
GenBank mRNA | AB174344.1 | - | 3 - 4 | 235 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
