Assay Details
| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284122.1 | NP_001271051.1 | 1 - 2 | 270 | 76 | |
| XM_005563056.2 | XP_005563113.1 | 1 - 2 | 286 | 76 | ||
| GenBank mRNA | AB179450.1 | - | 1 - 2 | 270 | 76 | |
More Information
Set Membership: |
|

BRF2
Genomic Map