Assay Details
| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284635.1 | NP_001271564.1 | 4 - 5 | 440 | 72 | |
| XM_005589356.2 | XP_005589413.1 | 4 - 5 | 778 | 72 | ||
| XM_005589359.2 | XP_005589416.1 | 3 - 4 | 705 | 72 | ||
| XM_005589360.2 | XP_005589417.1 | 3 - 4 | 428 | 72 | ||
| GenBank mRNA | AB169584.1 | - | 4 - 5 | 444 | 72 | |
| AB170222.1 | - | 4 - 5 | 436 | 72 | ||
| AB174290.1 | - | 4 - 5 | 440 | 72 | ||
More Information
Set Membership: |
|

DMAC2
Genomic Map