Assay Details
| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285364.1 | NP_001272293.1 | 2 - 3 | 1724 | 102 | |
| XM_015432091.1 | XP_015287577.1 | 3 - 4 | 1160 | 102 | ||
| XM_015432092.1 | XP_015287578.1 | 3 - 4 | 433 | 102 | ||
| XM_015432093.1 | XP_015287579.1 | 4 - 5 | 579 | 102 | ||
| XM_015432094.1 | XP_015287580.1 | 3 - 4 | 444 | 102 | ||
| GenBank mRNA | AB172141.1 | - | 2 - 3 | 1724 | 102 | |
More Information
Set Membership: |
|

ORMDL1
Genomic Map