Assay Details
Gene Symbol: TPD52L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283516.1 | NP_001270445.1 | 2 - 3 | 481 | 92 | |
GenBank mRNA | AB168902.1 | - | 2 - 3 | 481 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
