Assay Details
| Gene Symbol: STMN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283705.1 | NP_001270634.1 | 3 - 4 | 373 | 112 | |
| XM_015457625.1 | XP_015313111.1 | 3 - 4 | 542 | 112 | ||
| GenBank mRNA | AB169859.1 | - | 3 - 4 | 375 | 112 | |
More Information
Set Membership: |
|

STMN3
Genomic Map