Assay Details
| Gene Symbol: APOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283764.1 | NP_001270693.1 | 4 - 5 | 453 | 63 | |
| XM_005545330.1 | XP_005545387.1 | 4 - 5 | 577 | 63 | ||
| GenBank mRNA | AB072021.1 | - | 3 - 4 | 341 | 63 | |
| AB170247.1 | - | 4 - 5 | 453 | 63 | ||
More Information
Set Membership: |
|

APOD
Genomic Map