Assay Details
Gene Symbol: CRISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283995.1 | NP_001270924.1 | 3 - 4 | 210 | 68 | |
XM_005552768.1 | XP_005552825.1 | 4 - 5 | 319 | 68 | ||
XM_015448800.1 | XP_015304286.1 | 4 - 5 | 316 | 68 | ||
XM_015448801.1 | XP_015304287.1 | 4 - 5 | 296 | 68 | ||
XM_015448803.1 | XP_015304289.1 | 4 - 5 | 969 | 68 | ||
XM_015448804.1 | XP_015304290.1 | 3 - 4 | 264 | 68 | ||
XM_015448805.1 | XP_015304291.1 | 3 - 4 | 259 | 68 | ||
XM_015448806.1 | XP_015304292.1 | 3 - 4 | 932 | 68 | ||
GenBank mRNA | AB169287.1 | - | 3 - 4 | 210 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
