Assay Details
Gene Symbol: FAM118B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285380.1 | NP_001272309.1 | 3 - 4 | 411 | 67 | |
XM_005580120.2 | XP_005580177.1 | 3 - 4 | 455 | 67 | ||
XM_005580121.2 | XP_005580178.1 | 4 - 5 | 412 | 67 | ||
XM_005580122.2 | XP_005580179.1 | 3 - 4 | 652 | 67 | ||
XM_005580123.2 | XP_005580180.1 | 3 - 4 | 428 | 67 | ||
XM_005580125.2 | XP_005580182.1 | 3 - 4 | 455 | 67 | ||
XM_005580126.2 | XP_005580183.1 | 3 - 4 | 455 | 67 | ||
GenBank mRNA | AB168624.1 | - | 3 - 4 | 411 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
