Assay Details
Gene Symbol: PRRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283064.1 | NP_001269993.1 | 3 - 4 | 292 | 87 | |
XM_005593271.2 | XP_005593328.1 | 3 - 4 | 594 | 87 | ||
XM_005593273.2 | XP_005593330.1 | 3 - 4 | 351 | 87 | ||
GenBank mRNA | AB174198.1 | - | 3 - 4 | 292 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
