Assay Details
| Gene Symbol: PPP1R11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283316.1 | NP_001270245.1 | 1 - 2 | 290 | 78 | |
| XM_015448813.1 | XP_015304299.1 | 1 - 2 | 395 | 78 | ||
| GenBank mRNA | AB171371.1 | - | 1 - 2 | 290 | 78 | |
More Information
Set Membership: |
|

PPP1R11
Genomic Map