Assay Details
| Gene Symbol: CCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283464.1 | NP_001270393.1 | 6 - 7 | 1007 | 66 | |
| GenBank mRNA | AB168913.1 | - | 6 - 7 | 1007 | 66 | |
| AB169084.1 | - | 6 - 7 | 955 | 66 | ||
More Information
Set Membership: |
|

CCNB2
Genomic Map