Assay Details
| Gene Symbol: FAM122B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284691.1 | NP_001271620.1 | 5 - 6 | 458 | 65 | |
| XM_015443733.1 | XP_015299219.1 | 6 - 7 | 548 | 65 | ||
| XM_015443734.1 | XP_015299220.1 | 6 - 7 | 548 | 65 | ||
| XM_015443735.1 | XP_015299221.1 | 6 - 7 | 582 | 65 | ||
| XM_015443736.1 | XP_015299222.1 | 6 - 7 | 582 | 65 | ||
| XM_015443737.1 | XP_015299223.1 | 5 - 6 | 491 | 65 | ||
| XM_015443738.1 | XP_015299224.1 | 5 - 6 | 479 | 65 | ||
| XM_015443739.1 | XP_015299225.1 | 5 - 6 | 1309 | 65 | ||
| XM_015443741.1 | XP_015299227.1 | 6 - 7 | 548 | 65 | ||
| XM_015443742.1 | XP_015299228.1 | 6 - 7 | 548 | 65 | ||
| XM_015443743.1 | XP_015299229.1 | 6 - 7 | 582 | 65 | ||
| XM_015443744.1 | XP_015299230.1 | 6 - 7 | 536 | 65 | ||
| XM_015443745.1 | XP_015299231.1 | 5 - 6 | 479 | 65 | ||
| GenBank mRNA | AB172720.1 | - | 5 - 6 | 458 | 65 | |
More Information
Set Membership: |
|

FAM122B
Genomic Map