Assay Details
Gene Symbol: LRP2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285272.1 | NP_001272201.1 | 8 - 9 | 1185 | 61 | |
XM_005556422.2 | XP_005556479.1 | 8 - 9 | 1396 | 61 | ||
XM_005556423.2 | XP_005556480.1 | 8 - 9 | 1612 | 61 | ||
GenBank mRNA | AB179234.1 | - | 8 - 9 | 1185 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
