Assay Details
| Gene Symbol: S100A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283326.1 | NP_001270255.1 | 2 - 3 | 216 | 64 | |
| GenBank mRNA | AB173318.1 | - | 2 - 3 | 218 | 64 | |
More Information
Set Membership: |
|

S100A1
Genomic Map