Assay Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283903.1 | NP_001270832.1 | 3 - 4 | 1189 | 93 | |
XM_015433436.1 | XP_015288922.1 | 2 - 3 | 246 | 93 | ||
GenBank mRNA | AB174514.1 | - | 3 - 4 | 1189 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
