Assay Details
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283691.1 | NP_001270620.1 | 3 - 4 | 426 | 68 | |
XM_005562885.2 | XP_005562942.1 | 3 - 4 | 68 | |||
XM_015454368.1 | XP_015309854.1 | 3 - 4 | 68 | |||
GenBank mRNA | AB169623.1 | - | 3 - 4 | 455 | 68 | |
AB170441.1 | - | 3 - 4 | 426 | 68 | ||
AB171166.1 | - | 3 - 4 | 429 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
