Assay Details
| Gene Symbol: DDX19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284589.1 | NP_001271518.1 | 6 - 7 | 548 | 64 | |
| XM_015442711.1 | XP_015298197.1 | 7 - 8 | 682 | 64 | ||
| GenBank mRNA | AB169910.1 | - | 6 - 7 | 611 | 64 | |
| AB173768.1 | - | 6 - 7 | 548 | 64 | ||
More Information
Set Membership: |
|

DDX19A
Genomic Map