Assay Details
| Gene Symbol: SOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285406.1 | NP_001272335.1 | 1 - 2 | 70 | 87 | |
| GenBank mRNA | AB087270.1 | - | 1 - 2 | 70 | 87 | |
| AB220492.1 | - | 151 | 87 | |||
More Information
Set Membership: |
|

SOD1
Genomic Map