Assay Details
Gene Symbol: LOC102117143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283228.1 | NP_001270157.1 | 2 - 3 | 350 | 103 | |
GenBank mRNA | AM295861.1 | - | 2 - 3 | 315 | 103 | |
AM295862.1 | - | 2 - 3 | 315 | 103 | ||
AM295863.1 | - | 2 - 3 | 315 | 103 | ||
AM295864.1 | - | 2 - 3 | 315 | 103 | ||
AM295865.1 | - | 2 - 3 | 315 | 103 | ||
AM295866.1 | - | 2 - 3 | 315 | 103 | ||
AM295867.1 | - | 2 - 3 | 315 | 103 | ||
AM295868.1 | - | 2 - 3 | 315 | 103 | ||
AM295869.1 | - | 2 - 3 | 315 | 103 | ||
AM295870.1 | - | 2 - 3 | 315 | 103 | ||
AM295871.1 | - | 2 - 3 | 315 | 103 | ||
AM295872.1 | - | 2 - 3 | 315 | 103 | ||
AM295874.1 | - | 2 - 3 | 315 | 103 | ||
AM295875.1 | - | 2 - 3 | 315 | 103 | ||
AM295876.1 | - | 2 - 3 | 315 | 103 | ||
AM295877.1 | - | 2 - 3 | 315 | 103 | ||
EU203688.1 | - | 2 - 3 | 349 | 103 | ||
FJ430069.1 | - | 2 - 3 | 348 | 103 | ||
FJ430070.1 | - | 2 - 3 | 348 | 103 | ||
FM246485.1 | - | 2 - 3 | 315 | 103 | ||
GU063712.1 | - | 2 - 3 | 350 | 103 | ||
GU063713.1 | - | 2 - 3 | 350 | 103 | ||
GU063714.1 | - | 2 - 3 | 350 | 103 | ||
GU063715.1 | - | 2 - 3 | 350 | 103 | ||
HQ992789.1 | - | 1 - 2 | 275 | 103 | ||
LC043317.1 | - | 2 - 3 | 308 | 103 | ||
LC053847.1 | - | 2 - 3 | 314 | 103 | ||
LN851951.1 | - | 2 - 3 | 348 | 103 | ||
LN994432.1 | - | 2 - 3 | 348 | 103 | ||
LN994434.1 | - | 2 - 3 | 348 | 103 | ||
LN994436.1 | - | 2 - 3 | 348 | 103 | ||
LN994437.1 | - | 2 - 3 | 348 | 103 | ||
LN994438.1 | - | 2 - 3 | 348 | 103 | ||
LN994439.1 | - | 2 - 3 | 348 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
