Assay Details
Gene Symbol: CYP2C19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283290.1 | NP_001270219.1 | 5 - 6 | 848 | 94 | |
XM_015455918.1 | XP_015311404.1 | 5 - 6 | 670 | 94 | ||
GenBank mRNA | DQ074805.1 | - | 5 - 6 | 848 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
