Assay Details
| Gene Symbol: C1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284039.1 | NP_001270968.1 | 6 | 1016 | 121 | |
| GenBank mRNA | AB170629.1 | - | 6 | 1016 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1D| Gene Symbol: C1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284039.1 | NP_001270968.1 | 6 | 1016 | 121 | |
| GenBank mRNA | AB170629.1 | - | 6 | 1016 | 121 | |
Set Membership: |
|