Assay Details
| Gene Symbol: ELK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287616.1 | NP_001274545.1 | 2 | 618 | 90 | |
| XM_005593430.2 | XP_005593487.1 | 3 | 804 | 90 | ||
| GenBank mRNA | AB171665.1 | - | 2 | 620 | 90 | |
More Information
Set Membership: |
|

ELK1
Genomic Map