Assay Details
Gene Symbol: TFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548623.2 | XP_005548680.1 | 1 - 2 | 356 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
