Assay Details
| Gene Symbol: PLA2G2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441517.1 | XP_015297003.1 | 3 - 4 | 353 | 57 | |
| XM_015441522.1 | XP_015297008.1 | 3 - 4 | 353 | 57 | ||
More Information
Set Membership: |
|

PLA2G2C
Genomic Map