Assay Details
Gene Symbol: CCDC110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556431.1 | XP_005556488.1 | 4 - 5 | 301 | 101 | |
XM_005556432.1 | XP_005556489.1 | 4 - 5 | 301 | 101 | ||
XM_015451093.1 | XP_015306579.1 | 3 - 4 | 593 | 101 | ||
GenBank mRNA | AB069986.1 | - | 4 - 5 | 300 | 101 | |
AB070016.1 | - | 4 - 5 | 292 | 101 | ||
AB070100.1 | - | 4 - 5 | 269 | 101 | ||
AB071050.1 | - | 4 - 5 | 269 | 101 | ||
AB168213.1 | - | 4 - 5 | 300 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015451093.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
