Assay Details
Gene Symbol: CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005573255.2 | XP_005573312.1 | 5 - 6 | 618 | 94 | |
XM_005573256.2 | XP_005573313.1 | 5 - 6 | 618 | 94 | ||
XM_005573257.2 | XP_005573314.1 | 5 - 6 | 618 | 94 | ||
XM_005573258.2 | XP_005573315.1 | 5 - 6 | 618 | 94 | ||
XM_015432272.1 | XP_015287758.1 | 5 - 6 | 618 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
