Assay Details
Gene Symbol: LOC102116906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577515.2 | XP_005577572.1 | 2 - 3 | 204 | 89 | |
XM_005577516.2 | XP_005577573.1 | 2 - 3 | 220 | 89 | ||
XM_015434581.1 | XP_015290067.1 | 5 - 6 | 673 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
