Assay Details
Gene Symbol: C14H11orf95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434579.1 | XP_015290065.1 | 2 - 3 | 913 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C14H11orf95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434579.1 | XP_015290065.1 | 2 - 3 | 913 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |