Assay Details
| Gene Symbol: CD300LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584376.2 | XP_005584433.1 | 5 - 6 | 743 | 85 | |
| XM_005584377.2 | XP_005584434.1 | 4 - 5 | 641 | 85 | ||
| XM_005584382.2 | XP_005584439.1 | 4 - 5 | 698 | 85 | ||
| XM_015438552.1 | XP_015294038.1 | 5 - 6 | 896 | 85 | ||
| XM_015438553.1 | XP_015294039.1 | 4 - 5 | 794 | 85 | ||
| XM_015438555.1 | XP_015294041.1 | 5 - 6 | 963 | 85 | ||
More Information
Set Membership: |
|

CD300LG
Genomic Map