Assay Details
| Gene Symbol: NR2C2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588511.2 | XP_005588568.1 | 3 - 4 | 659 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR2C2AP| Gene Symbol: NR2C2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588511.2 | XP_005588568.1 | 3 - 4 | 659 | 73 | |
Set Membership: |
|