Assay Details
Gene Symbol: IGFLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588889.2 | XP_005588946.2 | 1 - 2 | 811 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
