Assay Details
Gene Symbol: C19H19orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589856.2 | XP_005589913.1 | 1 | 691 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C19H19orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589856.2 | XP_005589913.1 | 1 | 691 | 83 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |