Assay Details
| Gene Symbol: OPRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555138.1 | XP_005555195.1 | 2 - 3 | 223 | 78 | |
| XM_005555139.2 | XP_005555196.1 | 2 - 3 | 138 | 78 | ||
More Information
Set Membership: |
|

OPRPN
Genomic Map