Assay Details
Gene Symbol: KLLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565881.2 | XP_005565938.1 | 1 | 365 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KLLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565881.2 | XP_005565938.1 | 1 | 365 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |