Assay Details
| Gene Symbol: APOL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567456.2 | XP_005567513.1 | 2 - 3 | 352 | 64 | |
| XM_005567457.2 | XP_005567514.1 | 2 - 3 | 350 | 64 | ||
| XM_005567458.2 | XP_005567515.1 | 2 - 3 | 333 | 64 | ||
More Information
Set Membership: |
|

APOL6
Genomic Map