Assay Details
| Gene Symbol: WFDC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569159.2 | XP_005569216.1 | 1 - 2 | 232 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFDC10A| Gene Symbol: WFDC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569159.2 | XP_005569216.1 | 1 - 2 | 232 | 74 | |
Set Membership: |
|