Assay Details
Gene Symbol: OBP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015436458.1 | XP_015291944.1 | 2 - 3 | 256 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
